Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 Kč
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Czech Republic
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
SDIP/Europium set
SDIP/Europium set for membrane fusion assay
SDIP/Europium set for membrane fusion assay
SDIP/Europium set for membrane fusion assay
Catalog # 80105.
Supplier:
Biotium
CAS Number:
Expand image
SDIP/Europium set for membrane fusion assay
Catalog # 80105.
Supplier:
Biotium
CAS Number:
Specifications
Pk:
1 SET
Seq:
0125
Specifications
About this item
Product Details
:
Storage Temperature:
Refrigerator
Product Details