
3,3'-Di-n-hexyloxacarbocyanine iodide [DiOC6(3) iodide] membrane stain, for histology
Catalog # 70009.
Supplier: Biotium
CAS Number:
Specifications
- Pk:100 mg
- Seq:2440
Specifications
About this item
:
:572.531 g/mol
:GHS07
Storage Temperature:Refrigerator
CAS Number:53213-82-4
Formula:C₂₉H₃₇IN₂O₂
MDL Number:MFCD00011954