Order Entry
Czech Republic
ContactUsLinkComponent
3,3'-Diethyloxacarbocyanine iodide [DiOC2(3) iodide] membrane stain, for histology
3,3'-Diethyloxacarbocyanine iodide [DiOC2(3) iodide] membrane stain, for histology
Catalog # 70008.
Supplier:  Biotium
CAS Number:  
undefined
3,3'-Diethyloxacarbocyanine iodide [DiOC2(3) iodide] membrane stain, for histology
Catalog # 70008.
Supplier:  Biotium
CAS Number:  

Specifications

  • Pk:
    100 mg
  • Seq:
    2440

Specifications

About this item

 :
 :460.32 g/mol
Storage Temperature:Refrigerator
CAS Number:905-96-4
Formula:C₂₁H₂₁IN₂O₂
MDL Number:MFCD00011952