Supelco® Cerilliant® Organic Reference Standard, Vitamin K2-(MK-7)
Catalog # V-044-1ML
Supplier: Merck
CAS Number:
Specifications
- Concentration:100 µg/ml in acetonitrile
- Pk:1 mL
- Standards:Certified reference material, Cerilliant®
- CAS No.:2124-57-4
- Description:Vitamin K2-(MK-7)
- Storage conditions:–70 °C
- Hazard class description:H225 - H302 + H312 + H332 - H319
- Shipping conditions:WICE
- M-Clarity quality level:MQ300
- Synonym(s):(all-E)–2-(3,7,11,15,19,23,27-Heptamethyl-2,6,10,14,18,22,26-octacosaheptaenyl)-3-methyl-1,4-naphthalenedione
- Melting point:–48 °C
- Molecular weight:649,00
- Flash point:2,0 °C - Closed cup
Specifications
About this item
- GHS signal: GHS02, GHS07
Caution: P210 - P280 - P301 + P312 - P303 + P361 + P353 - P304 + P340 + P312 - P305 + P351 + P338