Order Entry
Czech Republic
ContactUsLinkComponent
pH and conductivity in seawater, analytical standard, Supelco®
pH and conductivity in seawater, analytical standard, Supelco®
Catalog # QC3146-500ML
Supplier:  Merck
CAS Number:  
pH and conductivity in seawater, analytical standard, Supelco®
Catalog # QC3146-500ML
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    500 mL
  • Description:
    pH and Conductivity in Seawater

Specifications

About this item