Supelco® Organic Reference Standard, ASTM® D6584 Monoolein solution
Catalog # CRM44893
Supplier: Merck
CAS Number:
Specifications
- Concentration:5000 µg/ml in pyridine
- Pk:1
- Standards:Certified reference material
- CAS No.:111-03-5
- Description:ASTM® D6584 Monoolein solution
- Storage conditions:2…8 °C
- Hazard class description:H225 - H302 + H312 + H332 - H315 - H319
- Shipping conditions:WICE
- M-Clarity quality level:MQ100
- Synonym(s):Monoolein solution
- Flash point:17,0 °C - Closed cup
Specifications
About this item
- GHS signal: GHS02, GHS07
Caution: P210 - P280 - P301 + P312 - P303 + P361 + P353 - P304 + P340 + P312 - P305 + P351 + P338