Order Entry
Czech Republic
Orders LinkContactUsLinkComponent
Supelco® Organic Reference Standard, ASTM® D6584 1,3-Diolein solution

Supelco® Organic Reference Standard, ASTM® D6584 1,3-Diolein solution

ASTM® D6584 1,3-Diolein solution, 5000 µg/ml in pyridine, Certified reference material, 1
Catalog # CRM44894
Supplier:   Merck
Supelco® Organic Reference Standard, ASTM® D6584 1,3-Diolein solution
Catalog # CRM44894
Supplier:  Merck
CAS Number:  
Supelco® Organic Reference Standard, ASTM® D6584 1,3-Diolein solution
Supelco® Organic Reference Standard, ASTM® D6584 1,3-Diolein solution

ASTM® D6584 1,3-Diolein solution, 5000 µg/ml in pyridine, Certified reference material, 1

CAS Number:  
Price
1 630,00 Kč
Log in for availability
1 * 1 KS
Product Details & Documents
  • GHS signal: GHS02, GHS07

Caution: P210 - P280 - P301 + P312 - P303 + P361 + P353 - P304 + P340 + P312 - P305 + P351 + P338

Specifications

Specifications

  • Concentration:
    5000 µg/ml in pyridine
  • Pk:
    1
  • Standards:
    Certified reference material
  • Description:
    ASTM® D6584 1,3-Diolein solution
  • Cat. No.:
    CRM44894
  • M-Clarity quality level:
    MQ100
  • Storage conditions:
    2…8 °C
  • Hazard class description:
    H225 - H302 + H312 + H332 - H315 - H319
  • MDL No.:
    MFCD00042913
  • Flash point:
    17,0 °C - Closed cup
  • Synonym(s):
    ASTM® D6584 1,3-Diolein Solution
  • CAS No.:
    2465-32-9
  • Shipping conditions:
    WICE

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.