
1,6-Diphenyl-1,3,5-hexatriene membrane stain, for histology
Catalog # 60021.
Supplier: Biotium
CAS Number:
Specifications
- Pk:100 mg
- Seq:2440
Specifications
About this item
:
:232.33 g/mol
:GHS07
Storage Temperature:Freezer
CAS Number:1720-32-7
Formula:(C₆H₅)CHCHCHCHCHCH(C₆H₅)
MDL Number:MFCD00004793