Order Entry
Czech Republic
ContactUsLinkComponent
 
Health
3,3'-Diaminobenzidine tetrahydrochloride (DAB HCl) kit peroxidase substrate
Catalog # 30015.
Supplier:  Biotium
CAS Number:  
Health
3,3'-Diaminobenzidine tetrahydrochloride (DAB HCl) kit peroxidase substrate
Catalog # 30015.
Supplier:  Biotium
CAS Number:  

Specifications

  • Pk:
    1 KIT
  • Seq:
    0075

Specifications

About this item

 :
 :360.11 g/mol
 :GHS08
Storage Temperature:Refrigerator
CAS Number:7411-49-6
Formula:C₁₂H₁₈Cl₄N₄
MDL Number:MFCD00012969