Order Entry
Czech Republic
ContactUsLinkComponent
Tetrabutylammonium dihydrogen phosphate for ion pair chromatography, Supelco®
Tetrabutylammonium dihydrogen phosphate for ion pair chromatography, Supelco®
Catalog # 86899-6X1AMP-F
Supplier:  Merck
CAS Number:  
undefined
Tetrabutylammonium dihydrogen phosphate for ion pair chromatography, Supelco®
Catalog # 86899-6X1AMP-F
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    1 KIT
  • Seq:
    0075
  • Pk Info:
    6× 1 ampoule

Specifications

About this item

 :
 :339.46 g/mol
CAS Number:5574-97-0
Formula:C₁₆H₃₈NO₄P
MDL Number:MFCD00064526