Pesticide standard, Fenthion, reference material, Honeywell Fluka™
HONCR1702-100MG
:
- Pack type:Glass Bottle
- Pk:100 mg
- Standards:Analytical Standard
- CAS No.:55-38-9
- Description:Fenthion
- Formula:C₁₀H₁₅O₃PS₂
- Storage temperature:2-8°C
- Description:Fenthion
- Form:Liquid
High quality organic reference materials produced according to ISO 9001.
: Supplied with complete Certificate of Analysis (CoA) with expiry date and lot specific analytical results.