Order Entry
Czech Republic
ContactUsLinkComponent
 
Pesticide standard, Quinoxyfen, reference material, Honeywell Fluka™
  HONCR1691-100MG
 :  
Pesticide standard, Quinoxyfen, reference material, Honeywell Fluka™
  HONCR1691-100MG
 :  

 

  • Pack type:
    Glass Bottle
  • Pk:
    100 mg
  • Standards:
    Analytical Standard
  • CAS No.:
    124495-18-7
  • Description:
    Quinoxyfen
  • Formula:
    C₁₅H₈Cl₂FNO
  • Storage temperature:
    Ambient
  • Description:
    Quinoxyfen
  • Form:
    Powder

 

 

High quality organic reference materials produced according to ISO 9001.

 : Supplied with complete Certificate of Analysis (CoA) with expiry date and lot specific analytical results.