Pesticide standard, Boscalid, reference material, Honeywell Fluka™
Catalog # HONCR1667-100MG
Supplier: Honeywell Chemicals
CAS Number:
Specifications
- Pack type:Glass Bottle
- Pk:100 mg
- Standards:Analytical Standard
- CAS No.:188425-85-6
- Description:Boscalid
- Formula:C₁₈H₁₂Cl₂N₂O
- Storage temperature:Ambient
- Description:Boscalid
- Form:Powder
Specifications
About this item
High quality organic reference materials produced according to ISO 9001.
Delivery: Supplied with complete Certificate of Analysis (CoA) with expiry date and lot specific analytical results.