Pesticide standard, Triflusulfuron-methyl, reference material, Honeywell Fluka™
Catalog # HONCR1666-100MG
Supplier: Honeywell Chemicals
CAS Number:
Specifications
- Pack type:Glass Bottle
- Pk:100 mg
- Standards:Analytical Standard
- CAS No.:126535-15-7
- Description:Triflusulfuron-methyl
- Formula:C₁₇H₁₉F₃N₆O₆S
- Storage temperature:Ambient
- Description:Triflusulfuron-methyl
- Form:Powder
Specifications
About this item
High quality organic reference materials produced according to ISO 9001.
Delivery: Supplied with complete Certificate of Analysis (CoA) with expiry date and lot specific analytical results.