Order Entry
Czech Republic
ContactUsLinkComponent
 
Pesticide standard, Methiocarb sulfone, reference material, Honeywell Fluka™
  HONCR1663-50MG
 :  
Pesticide standard, Methiocarb sulfone, reference material, Honeywell Fluka™
  HONCR1663-50MG
 :  

 

  • Pack type:
    Glass Bottle
  • Pk:
    50 mg
  • Standards:
    Analytical Standard
  • CAS No.:
    2179-25-1
  • Description:
    Methiocarb sulfone
  • Formula:
    C₁₁H₁₅NO₄S
  • Storage temperature:
    Ambient
  • Description:
    Methiocarb sulfone
  • Form:
    Crystalline

 

 

High quality organic reference materials produced according to ISO 9001.

 : Supplied with complete Certificate of Analysis (CoA) with expiry date and lot specific analytical results.