Pharmaceutical analytical standard, AHD-¹³C3, reference material, Honeywell Fluka™
Catalog # HONCR1658-10MG
Supplier: Honeywell Chemicals
CAS Number:
Specifications
- Pack type:Glass Bottle
- Pk:10 mg
- Standards:Analytical Standard
- CAS No.:1007476-86-9
- Description:AHD-¹³C3
- Formula:13C₃H₅N₃O₂
- Storage temperature:Ambient
- Description:AHD-¹³C3
- Form:Solid
Specifications
About this item
High quality organic reference materials produced according to ISO 9001.
Delivery: Supplied with complete Certificate of Analysis (CoA) with expiry date and lot specific analytical results.