Pharmaceutical analytical standard, AOZ-d4, reference material, Honeywell Fluka™
HONCR1651-10MG
:
- Pack type:Glass Bottle
- Pk:10 mg
- Standards:Analytical Standard
- CAS No.:1188331-23-8
- Description:AOZ-d4
- Formula:C₃H₂D₄N₂O₂
- Storage temperature:Ambient
- Description:AOZ-d4
- Form:Crystalline
High quality organic reference materials produced according to ISO 9001.
: Supplied with complete Certificate of Analysis (CoA) with expiry date and lot specific analytical results.