Order Entry
Czech Republic
ContactUsLinkComponent
Pesticide standard, Carbofuran-3-hydroxy, reference material, Honeywell Fluka™
Pesticide standard, Carbofuran-3-hydroxy, reference material, Honeywell Fluka™
Catalog # HONCR1648-10MG
CAS Number:  
Pesticide standard, Carbofuran-3-hydroxy, reference material, Honeywell Fluka™
Catalog # HONCR1648-10MG
CAS Number:  

Specifications

  • Pack type:
    Glass Bottle
  • Pk:
    10 mg
  • Standards:
    Analytical Standard
  • CAS No.:
    16655-82-6
  • Description:
    Carbofuran-3-hydroxy
  • Formula:
    C₁₂H₁₅NO₄
  • Storage temperature:
    -20°C
  • Description:
    Carbofuran-3-hydroxy
  • Form:
    Crystalline

Specifications

About this item

High quality organic reference materials produced according to ISO 9001.

Delivery: Supplied with complete Certificate of Analysis (CoA) with expiry date and lot specific analytical results.