Order Entry
Czech Republic
ContactUsLinkComponent
Pesticide standard, Fluopyram, reference material, Honeywell Fluka™
Pesticide standard, Fluopyram, reference material, Honeywell Fluka™
Catalog # HONCR1633-50MG
CAS Number:  
Pesticide standard, Fluopyram, reference material, Honeywell Fluka™
Catalog # HONCR1633-50MG
CAS Number:  

Specifications

  • Pack type:
    Glass Bottle
  • Pk:
    50 mg
  • Standards:
    Analytical Standard
  • CAS No.:
    658066-35-4
  • Description:
    Fluopyram
  • Formula:
    C₁₆H₁₁ClF₆N₂O
  • Storage temperature:
    Ambient
  • Description:
    Fluopyram
  • Form:
    Solid

Specifications

About this item

High quality organic reference materials produced according to ISO 9001.

Delivery: Supplied with complete Certificate of Analysis (CoA) with expiry date and lot specific analytical results.