Order Entry
Czech Republic
ContactUsLinkComponent
Organic analytical standards, 2-Ethoxyethanol, reference material, Honeywell Fluka™
Organic analytical standards, 2-Ethoxyethanol, reference material, Honeywell Fluka™
Catalog # HONCR1597-5ML
CAS Number:  
Organic analytical standards, 2-Ethoxyethanol, reference material, Honeywell Fluka™
Catalog # HONCR1597-5ML
CAS Number:  

Specifications

  • Pack type:
    Glass Bottle
  • Pk:
    5 mL
  • Standards:
    Analytical Standard
  • CAS No.:
    110-80-5
  • Description:
    2-Ethoxyethanol
  • Formula:
    C₂H₅OCH₂CH₂OH
  • Storage temperature:
    Ambient
  • Description:
    2-Ethoxyethanol
  • Form:
    Liquid

Specifications

About this item

High quality organic reference materials produced according to ISO 9001.

Delivery: Supplied with complete Certificate of Analysis (CoA) with expiry date and lot specific analytical results.