Tubing, silicone
Supplier: BURKLE
Certificates
About this item
Silicone tubing is a versatile transparent hose.
- Expands while using organic solvents
- Autoclavable, ethylene oxide
- Appropriate for food
- Withstand temperature from −60 to +180 °C
- Good resistance against weak acids
To the extend it is non-reactive to ionising solvents except chlorinated aliphatic and aromatic hydrocarbons.
Hardness (Shore A): 55 ±5
Caution: Should not use it for harsh solutions and strong acids.
Frequently Bought Together