Order Entry
Czech Republic
ContactUsLinkComponent
 
Supelco® Organic Reference Standard, F.A.M.E. Mix Glc-40
  1895-1AMP
 :  Merck
 :  
Supelco® Organic Reference Standard, F.A.M.E. Mix Glc-40
  1895-1AMP
 :  Merck
 :  

 

  • Pk:
    1 Ampoul
  • Standards:
    Certified reference material, ampule of 100 mg (Neat)
  • Description:
    F.A.M.E. Mix GLC-40
  • Storage conditions:
    –10...–25 °C
  • Shipping conditions:
    DICE
  • M-Clarity quality level:
    MQ100
  • Synonym(s):
    Fatty Acid Methyl Ester Mix
  • Pk Info:
    Ampoule of 100 mg

 

 

 : This certified reference material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025:2005.
 : 100 mg ampule contains approximately equal amounts by weight of: Palmitic acid methyl ester (C16:0) Stearic acid methyl ester (C18:0) Arachidic acid methyl ester (C20:0) Behenic acid methyl ester (C22:0)