Order Entry
Czech Republic
ContactUsLinkComponent
EPA TCL base/neutrals mix 1, certified reference material, Supelco®
EPA TCL base/neutrals mix 1, certified reference material, Supelco®
Catalog # 48900-U
Supplier:  Merck
CAS Number:  
EPA TCL base/neutrals mix 1, certified reference material, Supelco®
Catalog # 48900-U
Supplier:  Merck
CAS Number:  

Specifications

  • Concentration:
    2000 mug/mL each component in methylene chloride
  • Pk:
    1
  • Description:
    EPA TCL Base/Neutrals Mix 1
  • M-Clarity quality level:
    MQ100
  • Type:
    Certified reference material

Specifications

About this item