Order Entry
Czech Republic
ContactUsLinkComponent
Supelco® Organic Reference Standard, Polynuclear Aromatic Hydrocarbons 16 Solution
Supelco® Organic Reference Standard, Polynuclear Aromatic Hydrocarbons 16 Solution
Supplier:  Merck
Certificates
Supelco® Organic Reference Standard, Polynuclear Aromatic Hydrocarbons 16 Solution
Supplier:  Merck

Specifications

  • Storage conditions:
    2...30 °C
  • Hazard class description:
    H225 - H302 + H312 + H332 - H319 - H411
  • Shipping conditions:
    STD
  • M-Clarity quality level:
    MQ100
  • Concentration:
    In acetonitrile
  • Flash point:
    2 °C

Specifications

Frequently Bought Together