Order Entry
Czech Republic
ContactUsLinkComponent
 
EPA CLP Semi-Volatile calibration mix, certified reference material, Supelco®
Catalog # 506508.
Supplier:  Merck
CAS Number:  
EPA CLP Semi-Volatile calibration mix, certified reference material, Supelco®
Catalog # 506508.
Supplier:  Merck
CAS Number:  

Specifications

  • Concentration:
    1000 mug/mL each component in dichloromethane : benzene (3:1)
  • Pk:
    1
  • Description:
    EPA CLP Semivolatile Calibration Mix
  • Type:
    Certified reference material

Specifications

About this item