EPA 552 halogenated acetic acids mix, certified reference material, Supelco®
48047.
: Merck
:
- Concentration:2000 mug/mL each component in methyl tert-butyl ether
- Pk:1
- Description:EPA 552 Halogenated Acetic Acids Mix
- Type:Certified reference material