Order Entry
Czech Republic
ContactUsLinkComponent
EPA 552.2 acids calibration mix, certified reference material, Supelco®
EPA 552.2 acids calibration mix, certified reference material, Supelco®
Catalog # 47787.
Supplier:  Merck
CAS Number:  
EPA 552.2 acids calibration mix, certified reference material, Supelco®
Catalog # 47787.
Supplier:  Merck
CAS Number:  

Specifications

  • Concentration:
    in methyl tert-butyl ether (varied)
  • Pk:
    1
  • Description:
    EPA 552.2 Acids Calibration Mix
  • Type:
    Certified reference material

Specifications

About this item