EPA 8020/8240 Aromatic Volatiles Mix, certified reference material, Supelco®
Catalog # 47504.
Supplier: Merck
CAS Number:
Specifications
- Concentration:100 mug/mL each component in methanol
- Pk:1
- Description:EPA 8020/8240 Aromatic Volatiles Mix
- Type:Certified reference material