Order Entry
Czech Republic
ContactUsLinkComponent
EPA 8270 base/neutrals surrogate spike mix HC, certified reference material, Supelco®
EPA 8270 base/neutrals surrogate spike mix HC, certified reference material, Supelco®
Catalog # 47263.
Supplier:  Merck
CAS Number:  
EPA 8270 base/neutrals surrogate spike mix HC, certified reference material, Supelco®
Catalog # 47263.
Supplier:  Merck
CAS Number:  

Specifications

  • Concentration:
    5000 mug/mL each component in dichloromethane
  • Pk:
    1
  • Description:
    EPA 8270 Base/Neutrals Surrogate Spike Mix HC
  • Type:
    Certified reference material

Specifications

About this item