EPA Phthalate esters mix, certified reference material, Supelco®
Catalog # 48231.
Supplier: Merck
CAS Number:
Specifications
- Concentration:2000 mug/mL each component in hexane
- Pk:1
- Description:EPA Phthalate Esters Mix
- Type:Certified reference material