Order Entry
Czech Republic
ContactUsLinkComponent
 
Supelco® Organic Reference Standard, Polynuclear Aromatic Hydrocarbons 16 Solution
Catalog # 36991-2ML
Supplier:  Merck
CAS Number:  
Supelco® Organic Reference Standard, Polynuclear Aromatic Hydrocarbons 16 Solution
Catalog # 36991-2ML
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    2 mL
  • Standards:
    Analytical standard, in cyclohexane
  • Description:
    Polynuclear aromatic hydrocarbons 16 solution
  • Seq:
    02
  • Storage conditions:
    No temperature limit
  • Hazard class description:
    H225 - H304 - H315 - H336 - H410
  • Shipping conditions:
    STD
  • MDL No.:
    MFCD00148504
  • M-Clarity quality level:
    MQ100
  • Concentration:
    In cyclohexane
  • Flash point:
    –18 °C

Specifications

About this item

  • GHS signal: GHS02, GHS08, GHS07, GHS09
Caution: P210 - P233 - P273 - P301 + P310 - P303 + P361 + P353 - P331