Order Entry
Czech Republic
ContactUsLinkComponent
Organic reference standard, N,N'-1,6-Hexanediylbis(4-(2-pyridinyl)-1piperazinecarboxamide) solution (derivative of 1,6-HDIP), certified reference material, Supelco®
Organic reference standard, N,N'-1,6-Hexanediylbis(4-(2-pyridinyl)-1piperazinecarboxamide) solution (derivative of 1,6-HDIP), certified reference material, Supelco®
Catalog # 48146.
Supplier:  Merck
CAS Number:  
Organic reference standard, N,N'-1,6-Hexanediylbis(4-(2-pyridinyl)-1piperazinecarboxamide) solution (derivative of 1,6-HDIP), certified reference material, Supelco®
Catalog # 48146.
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    1
  • Standards:
    Certified reference material
  • CAS No.:
    72375-27-0
  • Description:
    N,N'-1,6-Hexanediylbis(4-(2-pyridinyl)-1piperazinecarboxamide) solution (derivative of 1,6-HDIP)
  • Formula:
    C₂₆H₃₈N₈O₂
  • Storage temperature:
    RT
  • Description:
    N,N'-1,6-Hexanediylbis(4-(2-pyridinyl)-1piperazinecarboxamide) solution (derivative of 1,6-HDIP)
  • Pk Info:
    1 ml/ampule

Specifications

About this item

Certifications: This certified reference material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025:2005.