Supelco® Organic Reference Standard, BDE No 15 Solution
Catalog # 33662-1ML
Supplier: Merck
CAS Number:
Specifications
- Pk:1 mL
- Standards:Analytical standard
- CAS No.:2050-47-7
- Description:BDE No 15 solution
- Storage conditions:2...8 °C
- Hazard class description:H225 - H304 - H315 - H336 - H410
- Shipping conditions:STD
- MDL No.:MFCD00000095
- M-Clarity quality level:MQ100
- Synonym(s):4,4′-Dibromodiphenyl ether solution; 4,4′-DiBDE; PBDE 15
- Formula:C₁₂H₈Br₂O
- Molecular weight:328,00
- Storage temperature:2...8 °C
- Flash point:–12 °C
Specifications
About this item
- GHS signal: GHS02, GHS08, GHS07, GHS09
Caution: P210 - P273 - P301 + P310 + P331 - P302 + P352