Order Entry
Czech Republic
ContactUsLinkComponent
Supelco® Organic Reference Standard, All-Cis-4,7,10,13,16,19-Docosahexaenoic Acid Methyl Ester
Supelco® Organic Reference Standard, All-Cis-4,7,10,13,16,19-Docosahexaenoic Acid Methyl Ester
Catalog # 05832-100MG
Supplier:  Merck
CAS Number:  
undefined
Supelco® Organic Reference Standard, All-Cis-4,7,10,13,16,19-Docosahexaenoic Acid Methyl Ester
Catalog # 05832-100MG
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    100 mg
  • Standards:
    Analytical standard
  • CAS No.:
    2566-90-7
  • Description:
    all-cis-4,7,10,13,16,19-Docosahexaenoic acid methyl ester
  • Storage conditions:
    –20 °C
  • Shipping conditions:
    STD
  • MDL No.:
    MFCD00673450
  • M-Clarity quality level:
    MQ100
  • Synonym(s):
    Fame 22:6n-3; Methyl DHA; Methyl all-cis-4,7,10,13,16,19-docosahexenoate
  • Formula:
    C₂₃H₃₄O₂
  • Molecular weight:
    342,51
  • Storage temperature:
    −20 °C
  • Flash point:
    Not applicable

Specifications

About this item

Packaging: Bottomless glass bottle. Contents are inside inserted fused cone.