Supelco® Organic Reference Standard, Alkylphenol Internal Standard Mix 7-Solution For Din En Iso 18857-2
Catalog # 33627-10ML
Supplier: Merck
CAS Number:
Specifications
- Pk:10 mL
- Standards:Analytical standard
- Description:Alkylphenol Internal Standard Mix 7-solution for DIN EN ISO 18857-2
- Storage conditions:–20 °C
- Hazard class description:H225 - H319 - H336
- Shipping conditions:STD
- M-Clarity quality level:MQ100
- Storage temperature:−20 °C
- Concentration:In acetone
- Flash point:–17 °C - closed cup
Specifications
About this item
- GHS signal: GHS02, GHS07
Caution: P210 - P233 - P240 - P241 - P242 - P305 + P351 + P338