Organic reference standard, 4-(3,6-Dimethyl-3-heptyl)phenol-3,5-d2 solution, analytical standard, Supelco®
Catalog # 32274-1ML
Supplier: Merck
CAS Number:
Specifications
- Concentration:100 µg/ml in acetone
- Pk:1 mL
- Standards:Analytical standard
- CAS No.:1173020-19-3
- Description:4-(3,6-Dimethyl-3-heptyl)phenol-3,5-d2 solution
- Formula:C₁₅D₂H₂₂O
- Storage temperature:−20 °C
- Description:4-(3,6-Dimethyl-3-heptyl)phenol-3,5-d2 solution