Order Entry
Czech Republic
ContactUsLinkComponent
Supelco® Organic Reference Standard, 1,3-Butadiene diepoxide
Supelco® Organic Reference Standard, 1,3-Butadiene diepoxide
Catalog # 90474-250MG
Supplier:  Merck
CAS Number:  
undefined
Supelco® Organic Reference Standard, 1,3-Butadiene diepoxide
Catalog # 90474-250MG
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    250 mg
  • Standards:
    Analytical standard
  • CAS No.:
    1464-53-5
  • Description:
    1,3-Butadiene diepoxide
  • Storage conditions:
    2…8 °C
  • Hazard class description:
    H226 - H301 - H310 + H330 - H314 - H340 - H350
  • Shipping conditions:
    STD
  • MDL No.:
    MFCD00005120
  • M-Clarity quality level:
    MQ100
  • Synonym(s):
    1,2,3,4-Diepoxybutane
  • Melting point:
    2 - 4 °C (lit.)
  • Molecular weight:
    86,09
  • Density:
    1,113 g/ml at 25 °C (lit.)
  • Flash point:
    46 °C - Closed cup
  • Boiling point:
    56 - 58 °C/25 mmHg (lit.)

Specifications

About this item

  • GHS signal: GHS02, GHS06, GHS08, GHS05
Caution: P202 - P210 - P280 - P303 + P361 + P353 - P304 + P340 + P310 - P305 + P351 + P338