ContactUsLinkComponent
Polyethylene glycol Standard ReadyCal Set Mp 200-1200000, for GPC, Supelco
Polyethylene glycol Standard ReadyCal Set Mp 200-1200000, for GPC, Supelco
Catalog # 02393-1EA
Supplier:  Merck
CAS Number:  
undefined
Polyethylene glycol Standard ReadyCal Set Mp 200-1200000, for GPC, Supelco
Catalog # 02393-1EA
Supplier:  Merck
CAS Number:  
Specifications
  • Pk:
    1
  • Description:
    ReadyCal Set Mp 200 - 1200000
  • M-Clarity quality level:
    MQ100
Specifications
About this item
Set contains 10 different polyethylene glycol/polyethylene oxide standards with Mp ~194, ~970, ~2010, ~4450, ~15000, ~22100, ~62000, ~217000, ~490000, ~1190000; always 3 or 4 standards are lyophilised in a 1,5 ml vial (black, blue or yellow); for 1 complete calibration use one of each of the black, blue and yellow vials; 1 set contains 30 vials for 10 complete calibrations.
Certifications: Exact values can be taken from the accompanying certificate of analysis
Caution: Storage temperature: 2 to 8 °C. Limited shelf life, expiry date on the label.