Order Entry
Czech Republic
ContactUsLinkComponent
Kovac's indole reagent for microbiology pH <1 (H₂O, 20 °C), Millipore®
Flammable
Corrosive
Irritant
Kovac's indole reagent for microbiology pH <1 (H₂O, 20 °C), Millipore®
Catalog # 1.09293.0100
Supplier:  Merck
CAS Number:  
Flammable
Corrosive
Irritant
Kovac's indole reagent for microbiology pH <1 (H₂O, 20 °C), Millipore®
Catalog # 1.09293.0100
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    100 mL
  • Seq:
    2440

Specifications

Specification Test Results

  • Indole formation (Escherichia coli ATCC 25922 (WDCM 00013)):
    +
  • Indole formation (Proteus vulgaris ATCC 8427):
    +
  • Indole formation (Enterobacter aerogenes ATCC 13048):
    -

Specification Test Results

About this item

 :
 :149.19 g/mol
ADR:8 (3),II
 :M-Clarity™ quality level = MQ100
UN:2920
 :GHS02
 :GHS05
 :GHS07
Storage Temperature:Refrigerator
CAS Number:100-10-7
Formula:(CH₃)₂NC₆H₄CHO
MDL Number:MFCD00003381