Sodium stearyl fumarate 99.0-101.5% (by anhydrous basis) NF
: Fumaric Acid Stearyl Ester Sodium Salt, Fumaric Acid Octadecyl Ester Sodium Salt, Sodium Octadecyl Fumarate
:4070-80-8
:
:C₂₂H₄₀O₄
:MFCD00236640
:390.54 g/mol
- Environmentally Preferable:
- Assay (Anhydrous Basis):99.0 - 101.5%
- Saponification Value:142.2 - 146.0
- Water Determination:Max. 5.0%
- Lead (Pb):Max. 10 ppm
- Heavy Metals:Max. 20 ppm
- Sodium Stearyl Maleate And Stearyl Alcohol:To Pass Test
- Residual Solvents:To Pass Test
Frequently Bought Together