Order Entry
Czech Republic
ContactUsLinkComponent
Absorption cuvettes with lids
Absorption cuvettes with lids
Catalog # HELLKS4C-10K01M40L
Supplier:  HELLMA
Absorption cuvettes with lids
Catalog # HELLKS4C-10K01M40L
Supplier:  HELLMA

Specifications

  • Path length:
    10 mm
  • Volume:
    1,4 ml
  • Description:
    Cell absorbance, 200 - 2500 nm with lid spectrally calibrated
  • Pk:
    1 PAIR

Specifications

About this item