Order Entry
Czech Republic
ContactUsLinkComponent
 
 
 
 
Methanol, anhydrous ≥99.8% (by GC), Macron Fine Chemicals™
    
 
undefined
 
 
 
Methanol, anhydrous ≥99.8% (by GC), Macron Fine Chemicals™
    
 

 

  • Environmentally Preferable:
    Safer Human and Environmental Health
  • Environmental certification:
    Chem21 Solvent Selection Guide recommended after discussion

 

 

  • Assay (CH₃OH) (by GC):
    ≥99.8 %
  • Water (H₂O)(by Karl Fischer titrn):
    ≤0.1 %
  • Substances Reducing KMnO₄:
    Passes Test
  • Acidity:
    Passes Test
  • Acetone:
    Passes Test
  • Solubility in H₂O:
    Passes Test

 

 

Frequently Bought Together