Zadání objednávky
Czech Republic
ContactUsLinkComponent
Manganese(II) nitrate solution 50 - 52%, BAKER ANALYZED®, J.T.Baker®
Oxidizing
Corrosive
Health
Irritant
Manganese(II) nitrate solution 50 - 52%, BAKER ANALYZED®, J.T.Baker®
Catalog # 2544-08
Supplier:  Avantor
CAS Number:  
Oxidizing
Corrosive
Health
Irritant
Manganese(II) nitrate solution 50 - 52%, BAKER ANALYZED®, J.T.Baker®
Catalog # 2544-08
Supplier:  Avantor
CAS Number:  

Specifications

  • Bal.:
    1
  • Sekvence:
    0001

Specifications

Specification Test Results

  • Assay (Mn(NO₃)₂):
    50.0 - 52.0%
  • Chloride (Cl):
    ≤0.002%
  • Sulphate (SO₄):
    ≤0.01%
  • Substances Not Precipitated by (NH₄)₂S (as SO₄):
    ≤0.2%
  • Other Heavy Metals (as Pb):
    ≤0.002%
  • Trace Impurities - Iron (Fe):
    ≤0.001%
  • Zinc (Zn) :
    ≤0.02%

Specification Test Results

About this item

 :
 :178.95
UN:3093
ADR:8,II
 :GHS03
 :GHS05
 :GHS08
 :GHS07
CAS Number:10377-66-9
Formula:Mn(NO₃)₂
MDL Number:MFCD00011112