Supelco® Organic Reference Standard, Dextran From Leuconostoc Mesenteroides
31416-100MG
: Merck
:
- Pk:100 mg
- Standards:Analytical standard, for GPC, Mw 1000
- CAS No.:9004-54-0
- Description:Dextran from Leuconostoc mesenteroides
- Seq:10
- Storage conditions:2...30 °C
- Shipping conditions:STD
- MDL No.:MFCD00130935
- M-Clarity quality level:MQ100
- Formula:(C₆H₁₀O₅)ₙ