Order Entry
Czech Republic
ContactUsLinkComponent
Test strips, qualitative, INDIPRO
Test strips, qualitative, INDIPRO
Catalog # MANA90765
Supplier:  MACHEREY-NAGEL
CAS Number:  
undefined
Test strips, qualitative, INDIPRO
Catalog # MANA90765
Supplier:  MACHEREY-NAGEL
CAS Number:  

Specifications

  • Chemical method:
    Protein proof with tetrabromophenol
  • Pk:
    60
  • Description:
    INDIPRO papers for protein contamination

Specifications

About this item

The test paper allows the detection of protein contamination on surfaces and tools coming in contact with food. The test kit consists of 60 test strips with reagents.

  • Colour change: Yellow to green

Limit of sensitivity: 50 µg BSA (bovine serum albumin)

Packaging: Pack of 60 test strips 10×95 mm and reagents