Pharmaceutical analytical standard, Promazine-d6 hydrochloride, reference material, Honeywell Fluka™
Catalog # HONCR1542-10MG
Supplier: Honeywell Chemicals
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:10 mg
- CAS No.:1246819-33-9
- Description:Promazine-d₆ hydrochloride
- Formula:C₁₇H₁₄D₆N₂S · HCl
- Storage temperature:Ambient
- Description:Promazine-d₆ hydrochloride
- Freezing point:169...172 °C
Specifications
About this item
High quality inorganic and organic reference materials produced according to ISO 9001.
- Ensure you have the information you need to quickly start your analysis, and pass relevant audits
- Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results