Pesticide reference standard, (R,S)-Metalaxyl Metabolite CGA 226048, neat, Honeywell Fluka™
Katalogové číslo HONCR1523-10MG
Supplier: Honeywell Chemicals
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:10 mg
- Description:(R,S)-Metalaxyl Metabolite CGA 226048
- Formula:C₁₈H₂₅NO₆
- Storage temperature:Ambient
- Description:(R,S)-Metalaxyl Metabolite CGA 226048
- Freezing point:68...70 °C
Specifications
About this item
High quality inorganic and organic reference materials produced according to ISO 9001.
- Ensure you have the information you need to quickly start your analysis, and pass relevant audits
- Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results