Order Entry
Czech Republic
ContactUsLinkComponent
Pesticide reference standard, Metsulfuron-methyl-d3, neat, Honeywell Fluka™
Pesticide reference standard, Metsulfuron-methyl-d3, neat, Honeywell Fluka™
Catalog # HONCR1520-10MG
CAS Number:  
Pesticide reference standard, Metsulfuron-methyl-d3, neat, Honeywell Fluka™
Catalog # HONCR1520-10MG
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    10 mg
  • Description:
    Metsulfuron-methyl-d₃
  • Formula:
    C₁₄H₁₂D₃N₅O₆S
  • Storage temperature:
    Ambient
  • Description:
    Metsulfuron-methyl-d₃

Specifications

About this item

High quality inorganic and organic reference materials produced according to ISO 9001.

  • Ensure you have the information you need to quickly start your analysis, and pass relevant audits
  • Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results