Order Entry
Czech Republic
ContactUsLinkComponent
 
Pharmaceutical analytical standard, Lidocaine-d10, reference material, Honeywell Fluka™
  HONCR1516-10MG
 :  
Pharmaceutical analytical standard, Lidocaine-d10, reference material, Honeywell Fluka™
  HONCR1516-10MG
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    10 mg
  • CAS No.:
    851528-09-1
  • Description:
    Lidocaine-d10
  • Formula:
    C₁₄H₁₂D₁₀N₂O
  • Storage temperature:
    Ambient
  • Description:
    Lidocaine-d10
  • Freezing point:
    186...189 °C

 

 

High quality inorganic and organic reference materials produced according to ISO 9001.

  • Ensure you have the information you need to quickly start your analysis, and pass relevant audits
  • Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results