Pesticide reference standard, Cyromazine-13C3, neat, Honeywell Fluka™
HONCR1504-10MG
:
- Pack type:Glass bottle
- Pk:10 mg
- CAS No.:1808990-94-4
- Description:Cyromazine-¹³C₃
- Formula:C₃¹³C₃H₁₀N₆
- Storage temperature:Ambient
- Description:Cyromazine-¹³C₃
- Freezing point:216...218 °C
High quality inorganic and organic reference materials produced according to ISO 9001.
- Ensure you have the information you need to quickly start your analysis, and pass relevant audits
- Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results